Learn More
PURUS™ Synthetic Cleanroom Paper
Cleanroom paper that excels in Tensile Strength. For writing, printing and photocopying in cleanroom environments, PURUS™ Cleanroom Paper provides excellent printability and heat toner adhesion and is recyclable.
€ 22.92 - € 340.00
Specifications
Autoclavable | Autoclavable |
---|---|
For Use With (Application) | For Class 10 to Class 1000 cleanrooms, writing, printing and photocopying in clean environments |
Plain paper | Yes |
Product Type | Cleanroom Paper |
Product Code | Brand | Color | Size | Weight (Metric) | Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Size | Weight (Metric) | Quantity | Price | Quantity & Availability | ||
16460041
|
PURUS™
LCIB 2048C |
Blue | A5 | 80 gsm | 250 Sheets/Pk., 20 Packs/Cs. |
€ 221.00 € 210.47 / Pack of 5000 Save € 10.53 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16470041
|
PURUS™
LCIG 2050C |
Green | A5 | 80 gsm | 250 Sheets/Pk., 20 Packs/Cs. |
€ 221.00 € 210.47 / Pack of 5000 Save € 10.53 4% Off |
Estimated Shipment: 24-11-2025
Log in to see stock. |
||
16480041
|
PURUS™
LCIK 2052C |
Pink | A5 | 80 gsm | 250 Sheets/Pk., 20 Packs/Cs. |
€ 221.00 € 210.47 / Pack of 5000 Save € 10.53 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16430051
|
PURUS™
LCIW 2046C |
White | A5 | 80 gsm | 250 Sheets/Pk., 20 Packs/Cs. |
€ 221.00 € 210.47 / Pack of 5000 Save € 10.53 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16410051
|
PURUS™
LCIW 2034C |
White | A5 | 100 gsm | 250 Sheets/Pk., 20 Packs/Cs. |
€ 265.00 € 252.96 / Pack of 5000 Save € 12.04 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
16440041
|
PURUS™
LCIB 2042C |
Blue | A5 | 100 gsm | 250 Sheets/Pk., 20 Packs/Cs. |
€ 273.00 € 252.96 / Pack of 5000 Save € 20.04 7% Off |
Estimated Shipment: 24-11-2025
Log in to see stock. |
||
15713579
|
PURUS™
LCIB 2025C |
Blue | A4 | 80 gsm | 250 Sheets/Pk., 10 Packs/Cs. |
€ 230.00 € 192.49 / Pack of 10 Save € 37.51 16% Off |
Estimated Shipment: 08-10-2025
Log in to see stock. |
||
15743579
|
PURUS™
LCIG 2017C |
Green | A4 | 80 gsm | 250 Sheets/Pk., 10 Packs/Cs. |
€ 224.00 € 192.49 / Pack of 10 Save € 31.51 14% Off |
Estimated Shipment: 03-10-2025
Log in to see stock. |
||
15753579
|
PURUS™
LCIK 2029C |
Pink | A4 | 80 gsm | 250 Sheets/Pk., 10 Packs/Cs. |
€ 237.00 € 183.51 / Pack of 10 Save € 53.49 22% Off |
Estimated Shipment: 13-11-2025
Log in to see stock. |
||
15773579
|
PURUS™
LCIW 2014C |
White | A4 | 80 gsm | 250 Sheets/Pk., 10 Packs/Cs. |
€ 224.00 € 192.49 / Pack of 10 Save € 31.51 14% Off |
Estimated Shipment: 03-10-2025
Log in to see stock. |
||
15783579
|
PURUS™
LCIW 2025C-4 |
White | A4 | 100 gsm | 250 Sheets/Pk., 10 Packs/Cs. | ||||
15793579
|
PURUS™
LCIY 2016C |
Yellow | A4 | 80 gsm | 250 Sheets/Pk., 10 Packs/Cs. |
€ 224.00 € 192.49 / Pack of 10 Save € 31.51 14% Off |
Estimated Shipment: 20-10-2025
Log in to see stock. |
||
15763579
|
PURUS™
LCIW 2030C |
White | A4 | 105 gsm | 250 Sheets/Pk., 10 Packs/Cs. |
€ 35.00 € 22.92 / Pack of 250 Save € 12.08 34% Off |
Estimated Shipment: 07-10-2025
Log in to see stock. |
||
16490041
|
PURUS™
LCIW 2031C |
White | A3 | 80 gsm | 250 Sheets/Pk., 5 Packs/Cs. |
€ 221.00 € 210.47 / Pack of 1250 Save € 10.53 4% Off |
Estimated Shipment: 20-11-2025
Log in to see stock. |
||
16400051
|
PURUS™
LCIW 2032C |
White | A3 | 100 gsm | 250 Sheets/Pk., 5 Packs/Cs. |
€ 265.00 € 252.96 / Pack of 1250 Save € 12.04 4% Off |
Estimated Shipment: 30-10-2025
Log in to see stock. |
||
16450041
|
PURUS™
LCIB 2044C |
Blue | A3 | 100 gsm | 250 Sheets/Pk., 5 Packs/Cs. |
€ 265.00 € 252.96 / Pack of 1250 Save € 12.04 4% Off |
Estimated Shipment: 30-10-2025
Log in to see stock. |
||
15723579
|
PURUS™
LCIB 2040C |
Blue | A3 | 80 gsm | 250 Sheets/Pk., 5 Packs/Cs. |
€ 340.00
Pack of 5 |
Estimated Shipment: 03-10-2025
Log in to see stock. |
||
Description
- Features high tensile strength and excellent heat resistance for enhanced copier and printing efficiency
- Proprietary binding technology results in very low levels of particles, nonvolatile residue (NVR) and ion extractables
- Unique precision cutting technology guarantees consistent sizing and cleanliness
- Produced in ISO 4 (Class 10) cleanroom
- Recyclable
- Suitable for use in ISO 4 to 5 (Class 10 to Class 1000) cleanrooms
Specifications
Autoclavable | |
For Class 10 to Class 1000 cleanrooms, writing, printing and photocopying in clean environments | |
Yes | |
Cleanroom Paper |