Learn More
PURUS™ Synthetic Cleanroom Paper
Cleanroom paper that excels in Tensile Strength. For writing, printing and photocopying in cleanroom environments, PURUS™ Cleanroom Paper provides excellent printability and heat toner adhesion and is recyclable.
Brand:  PURUS™ LCIB 2048C
 View more versions of this product
Product Code. 16460041
  • ONLINE EXCLUSIVE PRICE
    € 221.00
    € 210.47 / Pack of 5000
    Save € 10.53
    4% Off
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
  • Features high tensile strength and excellent heat resistance for enhanced copier and printing efficiency
  • Propriatary binding technology results in very low levels of particles, nonvolatile residue (NVR) and ion extractables
  • Unique precision cutting technology guarantees consistent sizing and cleanliness
  • Produced in ISO 4 (Class 10) cleanroom
  • Recyclable
  • Suitable for use in ISO 4 to 5 (Class 10 to Class 1000) cleanrooms
Specifications
Specifications
Synthetic
Blue
Autoclavable
Yes
250 Sheets/Pk., 20 Packs/Cs.
80 gsm
No
210 x 148 mm
For Class 10 to Class 1000 cleanrooms, writing, printing and photocopying in clean environments
Cleanroom Paper
A5
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.