Microcentrifuges
Thermo Scientific™ mySPIN™ 6 Mini Centrifuge
Thermo Scientific™ mySPIN™ 6 Mini Centrifuge
Our most compact centrifuge, the mySPIN 6 mini centrifuge, is ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microcentrifuge tubes.
Thermo Scientific™ Sorvall™ Legend™ Micro 17R Microcentrifuge
Thermo Scientific™ Sorvall™ Legend™ Micro 17R Microcentrifuge
The compact footprint of this microcentrifuge is ideal for routine laboratory work up to 17,000 x g and requiring refrigeration.
Corning™ Mini Microcentrifuge, 100-240V
Corning™ Mini Microcentrifuge, 100-240V
Corning™ Mini Microcentrifuge, 100-240V personal benchtop instrument operates at a fixed speed and is designed for quick spin downs of micro-samples.
Thermo Scientific™ mySPIN 12-minicentrifuge
Thermo Scientific™ mySPIN 12-minicentrifuge
Bring increased efficiency and user comfort to your lab with the mySPIN 12 mini centrifuge, ideal for microfilter cell separation and HPLC protocols, PCR, clinical and diagnostic applications, requiring fast spins (12,500rpm/9,800xg).
Labnet™ Prism™ Mini Centrifuge
Labnet™ Prism™ Mini Centrifuge
A compact and economical choice for centrifuges, allowing each workstation to be equipped with a personal centrifuge. Labnet™ Prism™ Mini Centrifuges are suitable for a wide range of molecular biology separations and quick spins.
Ohaus™ Frontier™ 5000 Series Mini Centrifuge
Ohaus™ Frontier™ 5000 Series Mini Centrifuge
Powerful and convenient mini-centrifuge for simple and quick life science separation applications.
Eppendorf™ Model 5430R microcentrifuges
Eppendorf™ Model 5430R microcentrifuges
Keep your samples cool with this refrigerated microcentrifuge. Eppendorf™ Model 5430R Microcentrifuges combine the best features of a microcentrifuge (small footprint) and multipurpose centrifuge (versatility) in one instrument.
Fisherbrand™ Centrifuge/Stirrer
Fisherbrand™ Centrifuge/Stirrer
The Fisherbrand™ Centrifuge / Stirrer is the industry's first high-speed centrifuge (12,500 rpm) and magnetic stirrer (3 L), combined into one unit. This design innovation enables a substantial cost savings for labs by not having to purchase two pieces of equipment, a stirrer and a centrifuge, because the Fisherbrand™ Centrifuge / Stirrer does it all.
Eppendorf™ MiniSpin™/MiniSpin™ plus Centrifuge
Eppendorf™ MiniSpin™/MiniSpin™ plus Centrifuge
With Eppendorf MiniSpin and MiniSpin plus centrifuges, there’s no need to share! These powerful, entry-level centrifuges are small enough that each workstation can have one. Key parts are made of metal for maximum safety, and the MiniSpin plus is fast enough for molecular biology separations.
Thermo Scientific™ Sorvall™ Legend™ Micro 21R Microcentrifuge
Thermo Scientific™ Sorvall™ Legend™ Micro 21R Microcentrifuge
Spin 24 x 2 mL at up to 21,100 x g with refrigeration to accelerate routine microcentrifuge tube preparation.
Eppendorf™ Centrifuge 5420
Eppendorf™ Centrifuge 5420
The Eppendorf Centrifuge 5420 offers 24-place capacity in a small footprint and supports nucleic acid purification and gel-extraction applications.
Eppendorf™ Model 5430 Microcentrifuges
Eppendorf™ Model 5430 Microcentrifuges
Unique crossover model combines microcentrifuge capability with sophisticated features and controls
Eppendorf™ Centrifuge 5425/5425 R
Eppendorf™ Centrifuge 5425/5425 R
Eppendorf’s newest 24-place microcentrifuges, Centrifuge 5425 and 5425 R, feature ergonomic operation and a large selection of rotors with a maximum speed of up to 21,300 x g, making them perfectly suited for all modern molecular biological applications in Eppendorf and PCR tubes from 0.2 to 5 mL.
Eppendorf™ Centrifuge 5418 R
Eppendorf™ Centrifuge 5418 R
The Eppendorf Centrifuge 5418 R is an 18-place centrifuge designed for low capacities using 1.5 – 2.0 mL tubes and Microtainers. Extremely compact with low access height, and very quiet operation it is capable of up to 16,873 x g, making it ideal for standard molecular biology applications.
Eppendorf™ Centrifuge 5910 Ri
Eppendorf™ Centrifuge 5910 Ri
Centrifuge 5910 Ri stands out with an intuitive touch screen interface and a range of innovative solutions for usability, documentation and user management. Its wide variety of fixed-angle and swing-bucket rotors accommodate a comprehensive spectrum of applications in a compact footprint.
Fisherbrand™ PCR Bundle
Fisherbrand™ PCR Bundle
Experience seamless PCR sampling, preparation, and storage with this specially curated PCR Bundle.
Thermo Scientific™ Sorvall™ Legend™ Micro 21 Microcentrifuge
Thermo Scientific™ Sorvall™ Legend™ Micro 21 Microcentrifuge
Support micro-volume protocols such as nucleic acid or protein lysate preparation and PCR reaction set-up, with microcentrifuge performance up to 21,100 x g.
Thermo Scientific™ Sorvall™ Legend™ Micro 17 Microcentrifuge
Thermo Scientific™ Sorvall™ Legend™ Micro 17 Microcentrifuge
Accelerate routine sample preparation up to 17,000 x g with generous capacity in a compact microcentrifuge footprint.
Fisherbrand™ accuSpin 21R Refrigerated Microcentrifuge
Fisherbrand™ accuSpin 21R Refrigerated Microcentrifuge
The Fisherbrand accuSpin 21 Microcentrifuges with a 24 x 1.5/2.0 mL rotor and a ClickSeal biocontainment lid that offers exceptional acceleration of up to 21,000 x g in as little as 12 seconds.
Eppendorf Centrifuge 5427 R
Eppendorf Centrifuge 5427 R
Centrifuge 5427 R contains a natural refrigerant (R290) to protect your samples and the environment. It features 9 different rotor options including an aerosol-tight 48-place rotor for high-throughput applications.
Eppendorf Centrifuge 5427 R (EU-IVD)
Eppendorf Centrifuge 5427 R (EU-IVD)
Centrifuge 5427 R contains a natural refrigerant (R290) to protect your samples and the environment. It features 9 different rotor options including an aerosol-tight 48-place rotor for high-throughput applications.
Fisherbrand™ Microplate Centrifuge
Fisherbrand™ Microplate Centrifuge
For fast spindowns of PCR microplates. The Fisherbrand Microplate Centrifuge has a small footprint with an easy-access removable rotor for easier cleaning.
Fisherbrand™ PlateFuge™ Microplate Microcentrifuge
Fisherbrand™ PlateFuge™ Microplate Microcentrifuge
The PlateFuge™ is the first microcentrifuge with a swing-out rotor for microplates and ensures that all reactants are in the bottom of the wells for proper concentrations and improved yields.
Fisherbrand™ accuSpin 21 Ventilated Microcentrifuge
Fisherbrand™ accuSpin 21 Ventilated Microcentrifuge
The Fisherbrand accuSpin 21 Microcentrifuges with a 24 x 1.5/2.0 mL rotor and a ClickSeal biocontainment lid that offers exceptional acceleration of up to 21,000 x g in as little as 12 seconds.
CORNING AXYGEN™ Axyspin™ Refrigerated Microcentrifuge
CORNING AXYGEN™ Axyspin™ Refrigerated Microcentrifuge
CORNING AXYGEN™ Axyspin™ Refrigerated Microcentrifuge is a high-performance microcentrifuge designed to quickly process samples.
OHAUS™ Frontier™ 5000 Series Micro Centrifuge
OHAUS™ Frontier™ 5000 Series Micro Centrifuge
OHAUS™ Frontier™ 5000 Series Micro Centrifuge provides high-speed operation for a range of essential lab applications. Engineered with convenient operation in mind, these centrifuges feature an intuitive interface, making all key parameters immediately accessible for observation or adjustment.
Gilson™ CENTRY™ 117 Microcentrifuge
Gilson™ CENTRY™ 117 Microcentrifuge
CENTRY™ 117 Microcentrifuge has an easy-to-use touchscreen and offers one-button recall of up to nine stored custom run programs. Its proprietary air-flow system ensures temperature stability, even on extended runs, preserving sample integrity. The rotor design accommodates either 1.5−2.0 mL tubes or PCR strips.
Gilson™ CENTRY™ 103 Minicentrifuge
Gilson™ CENTRY™ 103 Minicentrifuge
The CENTRY™ 103 Minicentrifuge is an easy-to-use and portable minicentrifuge, ideal for the quick separation of particles from a supernatant or for driving down of a drop of liquid from the walls of microtubes.
Benchmark™ myFuge™ 12 Microcentrifuge with COMBI-Rotor™
Benchmark™ myFuge™ 12 Microcentrifuge with COMBI-Rotor™
The myFuge™ 12 eliminates the need to change rotors when switching between microtubes and PCR strips. The included COMBI-Rotor™ accommodates 12 microtubes and 4 PCR strips, all at the same time.
  spinner