missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Color | Colorless |
---|---|
pH | 10 |
Physical Form | Liquid |
Solution Type | pH Buffer |
Formulation | 95 to 99% Deionized water, 0.1 to 0.5% Sodium hydroxide; caustic soda |
Odor | None |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10684863
![]() |
Mettler Toledo
51350038 |
250 mL | 1 Pk. |
26.65 EUR
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10422701
![]() |
Mettler Toledo
51350024 |
250 mL | 6 Pk. |
87.40 EUR
1500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Specifications
Colorless | |
Liquid | |
95 to 99% Deionized water, 0.1 to 0.5% Sodium hydroxide; caustic soda |
10 | |
pH Buffer | |
None |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title