missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 10, Mettler Toledo™
Shop All Mettler Toledo Products

Click to view available options
Quantity:
250 mL
Packaging:
1 Pk.
6 Pk.
Unit Size:
1500mL
250mL
Specifications
Specifications
| Color | Colorless |
| pH | 10 |
| Physical Form | Liquid |
| Quantity | 250 mL |
| Solution Type | pH Buffer |
| Formulation | 95 to 99% Deionized water, 0.1 to 0.5% Sodium hydroxide; caustic soda |
| Odor | None |
| Packaging | 1 Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido