missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 10, Mettler Toledo™
Shop All Mettler Toledo Products

Click to view available options
Quantity:
250 mL
Packaging:
1 Pk.
6 Pk.
Unit Size:
1500mL
250mL
Specifications
Specifications
| Color | Colorless |
| pH | 10 |
| Physical Form | Liquid |
| Quantity | 250 mL |
| Solution Type | pH Buffer |
| Odor | None |
| Packaging | 6 Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction