missing translation for 'onlineSavingsMsg'
Learn More

TWEEN 20, MP Biomedicals™

TWEEN 20, Molecular biology reagent, Viscous pale yellow liquid

602.23 DKK

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
Synonym Polyethyleneglycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate, Polysorbate 20, Polyoxyethylene (20) Sorbitan Monolaurate
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11417160
View Documents Promotion Details
MP Biomedicals
11TWEEN201
500 mL
ONLINE EXCLUSIVE PRICE
682.00 DKK
602.23 DKK / 500mL
Save 79.77 DKK
11% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • TWEEN 20 is a nonionic detergent widely used in biochemical applications, such as emulsifying agents for the preparation of stable oil-inwater emulsions.
  • It has been used in preextraction of membranes to remove peripheral proteins.
  • TWEEN 20 has been used as a blocking agent for membrane based immunoassays at a typical concentration of 0.05%.
  • TWEEN 20 can be used for lysing mammalian cells at a concentration of 0.05 to 0.5%. TWEEN 20 is a polyoxyethylene sorbitol ester.
Specifications

Chemical Identifiers

9005-64-5
522.68
HMFKFHLTUCJZJO-UHFFFAOYNA-N
2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
MFCD00165986
Polyethyleneglycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate, Polysorbate 20, Polyoxyethylene (20) Sorbitan Monolaurate
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Pale Yellow
Liquid
1.09 - 1.12(25°C)
< 1.33 hPa (< 1.00 mmHg, Lit.)
(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
≤3%
Polyethyleneglycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate, Polysorbate 20, Polyoxyethylene (20) Sorbitan Monolaurate
HMFKFHLTUCJZJO-UHFFFAOYNA-N
2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
Molecular Biology
9005-64-5
>100°C (literature)
>230°F/ >110°C (Lit.)
1.08 - 1.13
Brookfield Viscosity: 370-430 cps (25°C, neat, Lit.)
MFCD00165986
Soluble in Water(100mg/mL).
CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
522.68
Videos
Safety and Handling

Safety and Handling

missing translation for 'rtecsNumber' : TR7400000

missing translation for 'storageNote' : Store at Room temperature(15-30°C).

Product Identifier
  • Tween 20, molecular biology reagent

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.