Learn More
Epredia™ SuperFrost Plus™ Adhesion slides
Minimizes tissue loss during staining resulting in quick turn-around times and reduced risk associated with tissue loss and repeat testing. Epredia™ SuperFrost Plus Slides secure frozen and formalin-fixed, paraffin embedded tissue sections as well as cytology preparations.
Brand:  Epredia™ J7800AMNZ
187.82 DKK valid until 2025-07-04
Use promo code "24147" to get your promotional price.
 View more versions of this product
Product Code. 15438060
  • 278.00 DKK / Pack of 72
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Only null left
In Stock
Edge
Add to basket
This item is not returnable. View return policy
Description
Description
  • No time wasted on slide-coating procedures
  • No money wasted on expensive adhesives
  • Reliable and uniform coating
  • Minimal sample loss
  • Reliable and safe adhesive action
TRUSTED_SUSTAINABILITY
Specifications
Specifications
75 mm
90°, Ground
White
72/Pk.
25 mm
Adhesion Slide
Clipped
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.