Learn More
Cole-Parmer™ Block Heat Insert for Cole-Parmer™ BH-200 and BH-250 Series Block Heaters and CB-200 Series Chiller/Heater
Interchangeable heating blocks offer excellent thermal conductivity.
172.00€ - 505.00€
Specifications
Material Anodized Aluminum
Type Heating Block
View More Specs
Products 18
Product Code Brand Description Holds Price Quantity & Availability  
Product Code Brand Description Holds Price Quantity & Availability  
11593809
View Documents
Cole-Parmer™
36620-61
Blank with Thermometer Hole Tubes, Plates
181.00€
Each
Only null left
Estimated Shipment: 11-08-2025
Add to basket
11936498
View Documents Promotion Details
Cole-Parmer™
36610-50
Single Wide 12 Tubes, 17 x 47 mm (dia. x L)
172.00€
Each
Only null left
In Stock
Add to basket
11476588
View Documents Promotion Details
Cole-Parmer™
36610-63
Single Wide 80 x 0.2 mL Tube Strips
230.00€
Each
Only null left
Estimated Shipment: 11-08-2025
Add to basket
12942590
View Documents
Cole-Parmer™
36620-75
Gelation Timer Cup, Flat Bottom Tubes, Plates
505.00€
Each
Only null left
Estimated Shipment: 11-08-2025
Add to basket
15556413
View Documents Promotion Details
Cole-Parmer™
36620-92
Flat Bottom Wells 20 x 13 mm dia. Vials
192.00€
Each
Only null left
Estimated Shipment: 11-07-2025
Add to basket
12071602
View Documents
Cole-Parmer™
36620-65
Tapered Wells 20 x 1.5 mL Tubes
180.00€
Each
Only null left
In Stock
Add to basket
12081602
View Documents
Cole-Parmer™
36620-66
Tapered Wells 30 x 0.5 mL Tubes
182.00€
Each
Only null left
In Stock
Add to basket
12952590
View Documents Promotion Details
Cole-Parmer™
36620-82
Shallow, Conical 20 x 12 mm Vials
313.00€
Each
Only null left
Estimated Shipment: 11-08-2025
Add to basket
15294497
View Documents Promotion Details
Cole-Parmer™
36620-77
Conical Wells 5 x 50 mL Tubes
182.00€
Pack of 5
Only null left
In Stock
Add to basket
15319943
View Documents
Cole-Parmer™
36620-79
Conical Wells 5 x 30 mm dia. Tubes
182.00€
Each
Only null left
In Stock
Add to basket
11869564
View Documents Promotion Details
Cole-Parmer™
36620-63
Conical Wells 6 x 24 mm dia. Tubes/Vials
180.00€
Each
Only null left
Estimated Shipment: 11-08-2025
Add to basket
10129190
View Documents Promotion Details
Cole-Parmer™
36620-64
Conical Wells 6 x 26 mm dia. Tubes/Vials
180.00€
Each
Only null left
In Stock
Add to basket
11543809
View Documents
Cole-Parmer™
36620-56
Conical Wells 12 x 16 mm dia. Tubes/Vials
180.00€
Each
Only null left
In Stock
Add to basket
15218043
View Documents Promotion Details
Cole-Parmer™
36620-81
Conical Wells 8 x 20 mm dia. Tubes
182.00€
Each
Only null left
In Stock
Add to basket
12001612
View Documents Promotion Details
Cole-Parmer™
36620-71
Conical Wells 20 x 2 mL Tubes
180.00€
Each
Only null left
In Stock
Add to basket
11880705
View Documents
Cole-Parmer™
36620-52
Conical Wells 20 x 10 mm dia. Tubes/Vials
182.00€
Each
Only null left
In Stock
Add to basket
11523809
View Documents
Cole-Parmer™
36620-54
Conical Wells 20 x 13 mm dia. Tubes/Vials
180.00€
Each
Only null left
In Stock
Add to basket
11899694
View Documents Promotion Details
Cole-Parmer™
36620-51
Conical Wells 30 x 6 mm dia. Tubes/Vials
182.00€
Each
Only null left
Estimated Shipment: 11-08-2025
Add to basket
Description
Description
  • Versatile—interchangeable heating blocks accommodate various tubes and plates
  • Block heaters sold separately
Interchangeable aluminum heating blocks provide excellent thermal conductivity and accommodate various test tubes, cups, and plates. Depending on the combination of blocks selected, plastic spacers (36620-67) can be used to fill a half-width recess, ensuring optimal thermal performance. Most blocks come with a separate hole to accept a thermometer.
Order a flexi-block to accommodate multiple size tubes, vials, or cuvettes at once. Unskirted PCR plates can be heated in either size flexi-block. Skirted microtiter plates, deep-well plates, and PCR plates can only be incubated in the triple-width version.
Specifications
Specifications
Anodized Aluminum
Heating Block
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.