missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Implen™ NanoPhotometer™ Standard Solutions
Use to control the photometric accuracy of the NanoPhotometer™ NP120, NP80, N60 and N50 (NanoVolume)
Brand: Implen™ N-568-S
2233.02 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Standard solutions for the control of photometric accuracy
- Verifies the instrument's absorbance accuracy
- For use with NanoPhotometer™ NP120/NP80/N60/N50
- CAS-No: 877-24-7
Specifications
Non-toxic Potassium Hydrogen Phthalate Solution (PHP) | |
10 x 0.5 mL | |
Store at room temperature |
NanoPhotometer N60 and NP80 | |
10 Vials | |
280 nm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction