missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Implen™ NanoPhotometer™ Standard Solutions
Use to control the photometric accuracy of the NanoPhotometer™ NP120, NP80, N60 and N50 (NanoVolume)
€ 62.40 - € 321.00
Specifications
Type | Non-toxic Potassium Hydrogen Phthalate Solution (PHP) |
---|---|
Storage Requirements | Store at room temperature |
Wavelength | 280 nm |
Product Code | Brand | For Use With (Equipment) | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Packaging | Price | Quantity & Availability | ||||
15713217
|
Implen™
N-568-S2 |
NanoPhotometer N60 and NP80 | 2 x 0.5 mL |
€ 62.40
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15765805
|
Implen™
N-568-S |
NanoPhotometer N60 and NP80 | 10 x 0.5 mL |
€ 321.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16473909
|
Implen™
N-120-S |
NanoPhotometer N120 | 5 x 1 mL (Per Solution) |
€ 300.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Standard solutions for the control of photometric accuracy
- Verifies the instrument's absorbance accuracy
- For use with NanoPhotometer™ NP120/NP80/N60/N50
- CAS-No: 877-24-7
Specifications
Non-toxic Potassium Hydrogen Phthalate Solution (PHP) | |
Store at room temperature | |
280 nm |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title