missing translation for 'onlineSavingsMsg'
Learn More

Implen™ NanoPhotometer™ Standard Solutions

Use to control the photometric accuracy of the NanoPhotometer™ NP120, NP80, N60 and N50 (NanoVolume)

Brand:  Implen™ N-568-S2

 View more versions of this product

Product Code. 15713217

  • € 62.40 / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

  • Standard solutions for the control of photometric accuracy
  • Verifies the instrument's absorbance accuracy
  • For use with NanoPhotometer™ NP120/NP80/N60/N50
  • CAS-No: 877-24-7
Specifications

Specifications

Non-toxic Potassium Hydrogen Phthalate Solution (PHP)
2 x 0.5 mL
Store at room temperature
NanoPhotometer N60 and NP80
2 Vials
280 nm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only