Learn More
3M™ Secure Click™ Half Facepiece Reusable Respirator
3M™ Secure Click™ Half Facepiece Reusable Respirator is designed with smart and intuitive features, which makes it simple and reliable.
Brand:  3M™ 7100172003
 View more versions of this product
Product Code. 16456370
  • 53.10 € / Each
Only null left
Estimated Shipment: 04-09-2025
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy
Description
Description
  • Greater breathability and comfort provided by the world's first quad flow cartridge system.
  • Push button seal check helps bolster confidence that the respirator is properly sealed to the face.
  • Speaking diaphragm is designed to help provide easier communication while you work.
  • Confidence that filters and cartridges are installed properly provided by all new Secure Click™ connection.
  • Low profile half mask design offers a wide field of view and compatibility with certain welding and grinding shields.
  • Over-moulded design for easy assembly/disassembly.
  • Advanced silicone material for increased comfort and greater durability.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AS/NZS 1716:2012
Dark Gray
Half Facepiece Respirator
Without Speaking Diaphragm
10/Cs.
Secure Click™
Silicone, Nylon
Medium
HF-802
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.