Learn More
3M™ Secure Click™ Half Facepiece Reusable Respirator
Shop All 3M Products
Click to view available options
Size:
Medium
Large
Unit Size:
Each
Description
- Greater breathability and comfort provided by the world's first quad flow cartridge system.
- Push button seal check helps bolster confidence that the respirator is properly sealed to the face.
- Speaking diaphragm is designed to help provide easier communication while you work.
- Confidence that filters and cartridges are installed properly provided by all new Secure Click™ connection.
- Low profile half mask design offers a wide field of view and compatibility with certain welding and grinding shields.
- Over-moulded design for easy assembly/disassembly.
- Advanced silicone material for increased comfort and greater durability.
Specifications
Specifications
Certifications/Compliance | AS/NZS 1716:2012 |
Connection Type | Secure Click™ |
Color | Dark Gray |
Material | Silicone, Nylon |
Product Type | Half Facepiece Respirator |
Size | Medium |
Includes | Without Speaking Diaphragm |
Model | HF-802 |
Quantity | 10/Cs. |