missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Angular Beaker for use with Buerkle™ Telescopic Rod

The beaker is adaptable to different container shapes by adjustable angle setting (up to 90°).

Brand:  Buerkle™ 5354-0200

59.84 EUR valid until 2025-10-31
Use promo code "24158" to get your promotional price.


 View more versions of this product

Product Code. 10451183

  • ONLINE EXCLUSIVE PRICE
    € 92.60
    € 59.85 / Each
    Save € 32.75
    35% Off
View Available Promotions
Only null left
In Stock
Add to basket
This item is not returnable. View return policy

Description

Description

  • Polypropylene
  • For industry and water treatment
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Water Sampler
Not Autoclavable
2000 mL
For Industry and Water Treatment
Polypropylene
67.63 oz.
Angular
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.