Learn More
Grant Instruments™ Refrigerated Immersion Coolers
GREENER_CHOICE
Refrigerated immersion coolers consists of a cooling coil connected to a refrigeration unit by a flexible pipe. Extract heat continuously, with the bath control unit controlling temperature.
Brand:  Grant Instruments™ C1GR
 View more versions of this product
Product Code. 11466487
  • ONLINE EXCLUSIVE PRICE
    € 2905.00
    € 2829.00 / Each
    Save € 76.00
    2% Off
Only null left
Estimated Shipment: 20-10-2025
Add to basket
This item is not returnable. View return policy
Description
Description
Refrigerated immersion coolers consists of a cooling coil connected to a refrigeration unit by a flexible pipe. Extract heat continuously, with the bath control unit controlling temperature.
  • C1GR 0°C to 40°C
  • The cooling coil can be continuously immersed in liquids up to 100°C with the cooler switched off and may be used to cool liquid down from 100°C but it is not designed for continuous operation above 40°C.
  • C2GR: -15°C to 40°C
TRUSTED_SUSTAINABILITY
Specifications
Specifications
460 x 305 x 225 mm (D x W x H)
0°C to 40°C
120/230 V
350w at 20°C, 110w at 0°C
925 mm
120/230 V
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.