missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ QB Series Dry Block Heating Systems, Precision Digital

Provides excellent temperature control and a wide choice of interchangeable blocks for maximum flexibility. Grant Instruments™ QB Series Dry Block Heating Systems, Precision Digital combines superb temperature control and uniformity with high quality design and great versatility.

Brand:  Grant Instruments™ QBD1

5819.82 DKK valid until 2025-12-20
Use promo code "24121" to get your promotional price.


 View more versions of this product

Product Code. 11450958

  • 7115.00 DKK / Each
Equipment End of Year Sale
Use promo code 24121 at checkout
View Available Promotions
Only null left
Estimated Shipment: 07-10-2025
Add to basket
This item is not returnable. View return policy

Description

Description

Provides excellent temperature control and a wide choice of interchangeable blocks for maximum flexibility. Grant Instruments™ QB Series Dry Block Heating Systems, Precision Digital combines superb temperature control and uniformity with high quality design and great versatility.

  • High power heater for fast heat-up - 25°C to 100°C in only 15 minutes.
  • Simple-to-use rotor plus two keys provide access to the interactive interface for fast, accurate set-up and use
  • Clear digital display - easy to read from a distance for instant reassurance
  • Convenient timer facility, with audible buzzer, for reaction timing and function timing, e.g. delayed heater switch-on/turn-off
  • Wide range of interchangeable blocks enables you to mix and match sample tubes as desired - extraction tool supplied as standard for easy and safe removal of blocks.
  • Optional safety cover - protects samples from contamination and users from accidental contact with hot blocks
  • Overtemperature cut-out protects your samples and your workplace
  • High quality, robust construction in streamLined coolwall aluminum and chemical-resistant plastic - durable in demanding environments
  • Compact footprint to optimise use of benchspace
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aluminum
Ambient +5° to 130°C
1
0.1° at 37°C
240 mm
120 mm
120 or 230 V
Chemical Resistant Plastic
Dry Bath Block
0.1° at 37°C
1 to 999 mins
200 mm
230 mm
150 w
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.