Learn More
Buerkle™ Polypropylene Measuring Scoop
Scoops and measures powders and granules. Buerkle™ Polypropylene Measuring Scoops are calibrated to accurately assess volume at the brim.
Brand:  Buerkle™
 View more versions of this product
Product Code. 10524132
  • ONLINE EXCLUSIVE PRICE
    € 11.20
    € 9.22 / Each
    Save € 1.98
    17% Off
View Available Promotions
Only null left
In Stock
Add to basket
This item is not returnable. View return policy
Description
Description
Universal graduated shovel for filling and measuring powders and granules. For exact metering, the graduated shovel is to be overfilled and the excess at the upper edge of the shovel is to be wiped off.
  • White polypropylene construction
  • High chemical-resistance
  • Calibrated volume when full to the brim
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.