Learn More
Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators
Accommodates tubes of different diameters. Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators keeps test tubes secure during rotation
€ 69.00 - € 167.00
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
11411078
|
Grant Instruments™
PRS-10 |
10 Tubes up to 30 mm |
€ 97.55 € 70.00 / Each Save € 27.55 28% Off |
In Stock
Log in to see stock. |
|||||
11421028
|
Grant Instruments™
PRS-14 |
14 tubes up to 30mm |
€ 128.00 € 124.00 / Each Save € 4.00 3% Off |
In Stock
Log in to see stock. |
|||||
11431028
|
Grant Instruments™
PRS-22 |
22 Tubes up to 15 mm |
€ 96.35 € 94.00 / Each Save € 2.35 2% Off |
Estimated Shipment: 04-12-2025
Log in to see stock. |
|||||
11491068
|
Grant Instruments™
PRS-26 |
26 Tubes up to 15 mm, 1.5 -15 mL |
€ 107.00 € 104.00 / Each Save € 3.00 2% Off |
Estimated Shipment: 13-10-2025
Log in to see stock. |
|||||
11411028
|
Grant Instruments™
PRS-48 |
48 Tubes up to 15 mm |
€ 167.00
Each |
Estimated Shipment: 21-10-2025
Log in to see stock. |
|||||
10860891
|
Grant Instruments™
PRS-5-12 |
5 x 30 mm and 12 x 15 mm tubes |
€ 69.00
Each |
In Stock
Log in to see stock. ![]() |
|||||
12915511
|
Grant Instruments™
PRS-8-22 |
8 x 30 mm, 22 x 15 mm tubes |
€ 128.00 € 124.00 / Each Save € 4.00 3% Off |
Estimated Shipment: 04-12-2025
Log in to see stock. |
|||||
Description
- For use with PTR-35 and PTR-60 Series 360° vertical multi-function rotators
- Hold as few as 8 test tubes or as many as 48.
- Accommodates a variety of capacities