Learn More
Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators
Accommodates tubes of different diameters. Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators keeps test tubes secure during rotation
Brand:  Grant Instruments™ PRS-22
 View more versions of this product
Product Code. 11431028
  • ONLINE EXCLUSIVE PRICE
    € 96.35
    € 94.00 / Each
    Save € 2.35
    2% Off
Only null left
Estimated Shipment: 05-12-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • For use with PTR-35 and PTR-60 Series 360° vertical multi-function rotators
  • Hold as few as 8 test tubes or as many as 48.
  • Accommodates a variety of capacities
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTR-30
22 Tubes up to 15 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.