Learn More
Fisherbrand™ pH Indicator Paper Sticks
Measure the pH value quickly and easily directly at the point of interest. Fisherbrand™ pH indicator paper sticks are ready to use at any time and don't need any calibration, set-up or maintenance. Therefore, they are perfect in a variety of environments such as: chemical laboratories, industry, dairies, production, universities, schools and food processing.
73.03 DKK - 375.44 DKK
Specifications
Includes | Colour comparison chart |
---|---|
Quantity | 100 Sticks |
Type | pH Indicator Paper Stick |
Product Code | Brand | pH Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH Range | Price | Quantity & Availability | |||||
10642751
|
Fisherbrand™
92110.204231 |
0 to 14 |
375.44 DKK
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11966106
|
Fisherbrand™
92115 |
0 to 6 |
126.00 DKK 116.39 DKK / Pack of 100 Save 9.61 DKK 7% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10353641
|
Fisherbrand™
92130.204231 |
3.6 to 6.1 |
361.00 DKK
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10333501
|
Fisherbrand™
92120.204231 |
4.5 to 10 |
373.64 DKK
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11386454
|
Fisherbrand™
92150.204231 |
6 to 7.7 |
361.00 DKK 73.03 DKK / Pack of 100 Save 287.97 DKK 79% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11996056
|
Fisherbrand™
92125 |
7 to 14 |
120.00 DKK 116.39 DKK / Pack of 100 Save 3.61 DKK 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11346454
|
Fisherbrand™
92180.204231 |
0.3 to 2.3 |
361.00 DKK 73.79 DKK / Pack of 100 Save 287.21 DKK 79% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11316454
|
Fisherbrand™
92118.204231 |
2 to 9 |
361.00 DKK
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376454
|
Fisherbrand™
92140.204231 |
5.1 to 7.2 |
361.00 DKK
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15384184
|
Fisherbrand™
92140 |
5.1 to 7.2 |
120.00 DKK 116.39 DKK / 100 units Save 3.61 DKK 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396454
|
Fisherbrand™
92160.204231 |
7.5 to 9.5 |
361.00 DKK
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11360395
|
Fisherbrand™
92135 |
3.1 to 8.3 |
120.00 DKK 116.39 DKK / Pack of 100 Save 3.61 DKK 3% Off |
In Stock
Log in to see stock. |
|||||
10070552
|
Fisherbrand™
92122 |
6 to 10 |
120.00 DKK 116.39 DKK / Pack of 100 Save 3.61 DKK 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Measure the pH value quickly and easily directly at the point of interest. Fisherbrand™ pH indicator paper sticks are ready to use at any time and don't need any calibration, set-up or maintenance. Therefore, they are perfect in a variety of environments such as: chemical laboratories, industry, dairies, production, universities, schools and food processing.
85mm x 6mm sticks with different indicator papers sealed onto each strip. The pH is determined by comparison with a color chart supplied. The sticks are sufficiently long to avoid contact between the fingers and the test sample.- Provides outstanding measurements safely over the designated pH range
- Color scale on the test strip box allows exact comparison with the color developing on the test strip pad
- Indicator is covalently bonded on the pads and does not bleed into the sample – can be left in sample for an indefinite time without contamination
- Hydrophobic plastic handle protects user from sample and allows pH testing in narrow and deep vessels
- Available in a flat box so they fit into your pocket
Specifications
Colour comparison chart | |
100 Sticks | |
pH Indicator Paper Stick |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.