Learn More
Macherey-Nagel™ pH-Fix pH Indicator Strips
Highly appreciated standard strips for many pH testing applications.
Brand:  Macherey-Nagel™ 92190
 View more versions of this product
Product Code. 15324194
  • ONLINE EXCLUSIVE PRICE
    120.00 DKK
    116.39 DKK / 100 units
    Save 3.61 DKK
    3% Off
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Highly appreciated standard strips for many pH testing applications.
  • Easy dip and read procedure provides reliable results in seconds
  • Due to the patented pH-Fix technology, the indicator is chemically bound to the test pad
  • Dye does not wash out, so clothes stay clean and samples remain pure
Specifications
Specifications
1.7 to 3.8
pH Indicator Paper
6 mm
100 Strips
85 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.