missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ PELTOR™ LiteCom Headset
3M™ PELTOR™ LiteCom Headsets are hearing protectors with a built-in pre-programmed two-way communication radio and an integrated noise cancelling microphone. They’re built for demanding environments to help you communicate efficiently over hazardous noise.
Brand: 3M™ 7000108541
This item is not returnable.
View return policy
Description
- Hearing protector with built-in analogue two-way communication radio
- 8 channels in the PMR 446 MHz band
- 38 sub-channels
- Noise cancelling waterproof microphone provides voice-operated transmission (VOX) for hands free use
- Voice guided menu system
- Auto power off function
- Clear battery status indication
- Communication radio in the PMR 446 MHz frequency range.
- Use to help you communicate efficiently in high noise environments

Specifications
Communication Headset | |
Helmet Mounted | |
33 dB | |
CE Approved |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction