missing translation for 'onlineSavingsMsg'
Learn More

HYDRA-POINT™ Solvent G for two-component system, J.T. Baker™

Pyridine free solvent for volumetric Karl Fischer titration

753.00 DKK - 2150.00 DKK

Specifications

Use Test Suitability for Karl Fischer titration: Passes Test
UN Number 1992
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10211311
View Documents
JT Baker
8855.1000
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10189160
View Documents
JT Baker
8855.2500
2.5 L
2150.00 DKK
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Volumetric reagents for water determination by Karl Fischer titration
  • No matter what your requirements are for volumetric systems, HYDRA-POINT™ Karl Fischer reagents make water determination easy and trouble-free
  • They are pyridine-free and specially formulated for existing application procedures to give you consistent, reliable results for all types of samples
  • Formulated to reach each endpoint quickly, accurately and with reproducibility
  • Accuracy- reagents have titres that are more stable with less drift, helping you save time and reduce reagent usage
  • To be used with HYDRA-POINT™ Titrant 2 product no. 8845 or HYDRA-POINT™ Titrant 5 product no. 8844
Specifications

Specifications

Suitability for Karl Fischer titration: Passes Test
1992
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Hydra-Point Solvent G

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H361-Suspected of damaging fertility or the unborn child.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.

Supplemental information
  • MIXTURE LIST-Contains : methanol, imidazole,sulphur dioxide

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.