missing translation for 'onlineSavingsMsg'
Learn More

HYDRA-POINT™ Solvent G for two-component system, J.T. Baker™

Pyridine free solvent for volumetric Karl Fischer titration

Brand:  JT Baker 8855.2500


Quantity 2.5 L

 View more versions of this product

Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 10189160

  • 2150.00 DKK / 2.50 L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

  • Volumetric reagents for water determination by Karl Fischer titration
  • No matter what your requirements are for volumetric systems, HYDRA-POINT™ Karl Fischer reagents make water determination easy and trouble-free
  • They are pyridine-free and specially formulated for existing application procedures to give you consistent, reliable results for all types of samples
  • Formulated to reach each endpoint quickly, accurately and with reproducibility
  • Accuracy- reagents have titres that are more stable with less drift, helping you save time and reduce reagent usage
  • To be used with HYDRA-POINT™ Titrant 2 product no. 8845 or HYDRA-POINT™ Titrant 5 product no. 8844
Specifications

Specifications

2.5 L
Suitability for Karl Fischer titration: Passes Test
1992
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H361-Suspected of damaging fertility or the unborn child.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.