Learn More
SCAT™ HPLC Supply and Waste Set, V2.0, Suitable for All HPLC Systems
The all-round carefree package suitable for all HPLC and UHPLC systems.
5630.00 DKK - 7010.00 DKK
Specifications
For Use With (Equipment) Any HPLC System
View More Specs
Products 4
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
16458190
View Documents
SCAT™
307337
HPLC Supply and Waste Set, V2.0, suitable for all HPLC systems
5630.00 DKK
Each
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 17-09-2025
Add to basket
17865704
View Documents
SCAT™
307447
6140.00 DKK
1 set
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 11-09-2025
Add to basket
18626478
View Documents
SCAT™
307338
HPLC Supply and Disposal Set (12 Month), Non-Electrostatic Conductive
6240.00 DKK
1 set
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 09-09-2025
Add to basket
18636478
View Documents
SCAT™
307448
HPLC Supply and Disposal Set (12 Month), Electrostatic Conductive
7010.00 DKK
1 sheet
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 09-09-2025
Add to basket
Description
Description
  • Protects your system from contamination
  • Keeps solvent mixtures stable
  • High performance materials with excellent chemical resistance
The safety set connects tubing safely and blocks hazardous vapours. Air valves and exhaust filters provide safe pressure equalisation without under- or overpressure in solvent bottles and waste containers
Specifications
Specifications
Any HPLC System
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.