Learn More
SCAT™ HPLC Supply and Waste Set, V2.0, Suitable for All HPLC Systems
The all-round carefree package suitable for all HPLC and UHPLC systems.
Brand:  SCAT™ 307338
 View more versions of this product
Product Code. 18626478
  • ONLINE EXCLUSIVE PRICE
    6240.00 DKK
    4573.57 DKK / 1 set
    Save 1666.43 DKK
    26% Off
Only null left
Estimated Shipment: 02-10-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • Protects your system from contamination
  • Keeps solvent mixtures stable
  • High performance materials with excellent chemical resistance
The safety set connects tubing safely and blocks hazardous vapours. Air valves and exhaust filters provide safe pressure equalisation without under- or overpressure in solvent bottles and waste containers
Specifications
Specifications
Liquid Chromatography
4 x Safety Cap III, GL45, 8 x air valve, 1 x Safety Waste Cap, GL45, 1 x Exhaust filter L, V3.0, with splash guard and change indicator, service life 12 months, 1 x 5 litre canister, PE-HD, 4 x Laboratory bottle DURAN, GL45, 1000 ml, clear glass and round, with cap and pouring ring , 5 x PFA-fitting with integrated ferrule, 2.3 mm OD, violet, 5 x PFA fitting with integrated ferrule, 1.6 mm OD, green, 10 x PFA fitting with integrated ferrule, 3.2 mm OD, blue, 10 x PFA blind plug, 2 x connector, straight, 6 - 8 mm ID, 1 x connector, curved, 5.0 - 11.5mm ID, 1 x connector, angled, 9.5 - 10 mm ID
Complete Solvent Collection System
Any HPLC System
4
Non-Electrostatic Conductive
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.