missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ Safety Funnel 'ARNOLD' with Ball and Lid, V2.0

The strong safety funnel ARNOLD V2.0 with lid and ball meets the highest requirements for safe disposal of hazardous liquid waste due to its combination of two independent safety mechanisms.

Brand:  SCAT™ 317649

1193.52 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 17192245

  • 1475.00 DKK / Each
Only null left
Estimated Shipment: 10-09-2025
Add to basket
This item is not returnable. View return policy

Description

Description

The safety funnel ARNOLD V2.0 with lid and ball valve is available for different container threads. In order to be able to produce in an environmentally friendly and economical way, we supply individual variants with a thread adapter, which has no influence on the functionality or safety of the funnel.

  • Two safety features consist of a ball integrated in the funnel basin and a sealing ring incorporated in the hinged lid of the funnel, which prevent the escape of toxic or explosive gases
  • Made of high quality HDPE material for maximum stability and durability
  • Hinged lid with sealing ring for closure when the funnel is not in use
  • Optimized sieve for catching larger particles and for depositing laboratory bottles
  • Improved splash guard with integrated air channels to prevent bubble formation
  • Ball valve with automatic opening mechanism during filling
  • Individually shortenable safety lance for clean drainage of liquid waste
  • All essential components are removable for easy cleaning
Specifications

Specifications

200 mm (Funnel)
Not Autoclavable
Safe Disposal of Liquid and Volatile Wastes
Lance (220 mm), Lid, Ball Valve, Splash Guard, Removable Sieve
Safety Funnel
HDPE
Blue, White
Disposal Containers
S50
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.