missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ BH-250 Series Dry-Block Heater

Choose a heater, then blocks to suit your application needs.

Brand:  Cole-Parmer™ 36620-02

 View more versions of this product

Product Code. 15245607

  • 2065.00 EUR / Each
Only null left
In Stock
Add to basket
This item is not returnable. View return policy

Description

Description

  • Precision-engineered heaters with excellent temperature accuracy
  • Versatile range of interchangeable heating blocks work with many size sample tubes and plates
  • Take up less storage space—lightweight and flat-top surface allow units to be stacked when not in use

Use these heaters as a dry, constant temperature source for various lab and research applications. All models have independent indicator lights to show main power, heater action, and over-temperature cut out. The ''number of blocks accepted'' on the ordering table indicate the number of single-width blocks accepted by each block heater. A heater that accepts two single-width blocks will accept one double-width block. Use three- and four-block models for high-throughput applications such as chemical oxygen demand (COD).

Specifications

Specifications

Digital, Twin Control, 100°C
Heating Block
230 V
Ambient +5° to 100°C
2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.