missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™ 58-530W Series Nitrile Immersion Gloves

Recommended for outdoor applications. Enables users to handle wet or oily objects with less grip force and more control. The patented Ansell Grip TechnologyTM ensures that the coating polymer does not penetrate the liner during manufacture to prevent possible chemical leakage onto the skin.

410.18 DKK

Specifications

Back Material Nitrile
Cuff Material Nitrile
Applications Industrial
Coating Material Nitrile
Color Green, Gray
View More Specs

Products 5
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12649741
View Documents Promotion Details
Ansell™
58535W100
10
ONLINE EXCLUSIVE PRICE
623.00 DKK
410.18 DKK / 6 pairs
Save 212.82 DKK
34% Off
View alternative products
Save Up to 
»
Only null left
In Stock
Edge
Add to basket
 
12659741
View Documents Promotion Details
Ansell™
58535W110
11
ONLINE EXCLUSIVE PRICE
623.00 DKK
410.18 DKK / 6 pairs
Save 212.82 DKK
34% Off
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 10-03-2026
Add to basket
 
12669741
View Documents Promotion Details
Ansell™
58535W070
7
ONLINE EXCLUSIVE PRICE
623.00 DKK
410.18 DKK / 6 pairs
Save 212.82 DKK
34% Off
View alternative products
Save Up to 
»
Only null left
Estimated Shipment: 18-11-2025
Add to basket
 
12679741
View Documents Promotion Details
Ansell™
58535W080
8
ONLINE EXCLUSIVE PRICE
623.00 DKK
410.18 DKK / 6 pairs
Save 212.82 DKK
34% Off
View alternative products
Save Up to 
»
Only null left
In Stock
Edge
Add to basket
 
12689741
View Documents Promotion Details
Ansell™
58535W090
9
ONLINE EXCLUSIVE PRICE
623.00 DKK
410.18 DKK / 6 pairs
Save 212.82 DKK
34% Off
View alternative products
Save Up to 
»
Only null left
In Stock
Edge
Add to basket
 
Description

Description

  • Nitrile polymer coating reduces chemical leakage onto skin
  • For indoor applications
  • Nylon liner
  • Gauntlet cuff
  • Sea green shell, anthracite gray dip color
Specifications

Specifications

Nitrile
Industrial
Green, Gray
Nitrile
Gauntlet
White Nylon
Chemical resistance
Nitrile
Nitrile
Yes
Immersion Gloves
Nylon
6 Pairs
58-535W
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.