missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™ 58-530W Series Nitrile Immersion Gloves

Recommended for outdoor applications. Enables users to handle wet or oily objects with less grip force and more control. The patented Ansell Grip TechnologyTM ensures that the coating polymer does not penetrate the liner during manufacture to prevent possible chemical leakage onto the skin.

Brand:  Ansell™ 58535W090

348.64 DKK valid until 2025-12-31
Use promo code "24116" to get your promotional price.


 View more versions of this product

Product Code. 12689741

  • ONLINE EXCLUSIVE PRICE
    623.00 DKK
    410.18 DKK / 6 pairs
    Save 212.82 DKK
    34% Off
View Available Promotions
Only null left
In Stock
Edge
Add to basket

Description

Description

  • Nitrile polymer coating reduces chemical leakage onto skin
  • For indoor applications
  • Nylon liner
  • Gauntlet cuff
  • Sea green shell, anthracite gray dip color
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nitrile
Industrial
Green, Gray
Nitrile
9
Nylon
6 Pairs
58-535W
Nitrile
Nitrile
Yes
Immersion Gloves
Gauntlet
White Nylon
Chemical resistance
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.