missing translation for 'onlineSavingsMsg'
Learn More

HALYARD™ AERO CHROME™ High-Performance Surgical Gown

High-Performance Surgical Gown combines comfort, softness and breathability with high level protection against pathogens and fluids, including chemotherapy drugs.

207.67€ - 269.88€

Specifications

Product Type Surgical Gown
No. of Pockets None
Material Cool Shield Copolymer
Color Silver
Closure Type Hook and Loop Neck with Waist Ties
View More Specs

Products 12
Product Code Brand Size Sterility Price Quantity & Availability  
Product Code Brand Size Sterility Price Quantity & Availability  
30274682
View Documents
HALYARD™
48392
Large Non-sterile
234.29€
Pack of 40
Only null left
Estimated Shipment: 04-07-2025
Add to basket
 
30274725
View Documents
HALYARD™
48393
X-Large Non-sterile
207.67€
Pack of 32
Only null left
Estimated Shipment: 04-07-2025
Add to basket
 
30274736
View Documents
HALYARD™
48394
2X-Large Non-sterile
214.20€
Pack of 32
Only null left
Estimated Shipment: 04-07-2025
Add to basket
 
30274651
View Documents
HALYARD™
48395
Large Sterile
238.47€
Pack of 30
Only null left
Estimated Shipment: 04-07-2025
Add to basket
 
30274705
View Documents
HALYARD™
48396
X-Large Sterile
240.00€
Pack of 28
Only null left
Estimated Shipment: 04-07-2025
Add to basket
 
30274656
View Documents
HALYARD™
48397
Large, X-Long Sterile
251.02€
Pack of 30
Only null left
Estimated Shipment: 04-07-2025
Add to basket
 
30274668
View Documents
HALYARD™
48398
X-Large, X-Long Sterile
269.88€
Pack of 30
Only null left
Estimated Shipment: 04-07-2025
Add to basket
 
30274657
View Documents
HALYARD™
48399
Large, X-Long Non-sterile
226.29€
Pack of 36
Only null left
Estimated Shipment: 04-07-2025
Add to basket
 
30274722
View Documents
HALYARD™
48400
X-Large, X-Long Non-sterile
220.73€
Pack of 32
Only null left
Estimated Shipment: 04-07-2025
Add to basket
 
30274767
View Documents
HALYARD™
48401
2X-Large, X-Long Non-sterile
213.37€
Pack of 30
Only null left
Estimated Shipment: 04-07-2025
Add to basket
 
30274704
View Documents
HALYARD™
48402
2X-Large, X-Long Sterile
269.71€
Pack of 28
Only null left
Estimated Shipment: 04-07-2025
Add to basket
 
30274695
View Documents
HALYARD™
48403
2X-Large Sterile
257.71€
Pack of 28
Only null left
Estimated Shipment: 04-07-2025
Add to basket
 
Description

Description

  • Culmination of HALYARD™'s expertise in designing high-performance garments for critical applications in Healthcare, the AERO CHROME™ gowns offer an exceptional combination of Protection, Breathability, and Comfort.
  • Red collar: Easily identify a high-performance gown
  • Breathable back panel: maximizes airflow for cool comfort
  • Soft, light, and breathable fabric: keeps staff protected yet cool and comfortable
  • Low linting: helps reduce the risk of wound contamination and infection
  • Meets the High Performance per EN13795-1:2019: ensures superior protection
  • Meets AAMI Level 4 standard: provides the highest level of fluid and microbial protection in the critical zones
  • No breakthrough detected after 240 minutes of exposure to 52 chemotherapy drugs
  • Raglan Sleeves for extra comfort and free movements
  • Available Sterile and Non Sterile
  • Sterile gown packed individually and with 2 towels for added convenience
  • Non Sterile gowns packed bulk in a bag to reduce packaging
Specifications

Specifications

Surgical Gown
Cool Shield Copolymer
Hook and Loop Neck with Waist Ties
Knit
Raglan Sleeve
Not Autoclavable
None
Silver
Single-use
Yes
Regulation (EU) 2017/745 Class I, EN 13795, AAMI level 4, ASTM D6978-05
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.