Learn More
Cytiva AcroPak™ 300 Capsule with PTFE Membrane
AcroPak 300 Capsule offers broad chemical compatibility with PTFE membrane and polypropylene housing.
322.00 € - 8500.00 €
Specifications
Material (Membrane) PTFE
Pore Size 0.2 μm
Sterility Non-sterile
Filtration Area 280 cm2
Outlet Style Stepped Hose Barb
View More Specs
Products 2
Product Code Brand No. per Pack Price Quantity & Availability  
Product Code Brand No. per Pack Price Quantity & Availability  
17114221
View Documents Promotion Details
Cytiva
12082
3/Pk.
322.00 €
Pack of 3
Please to purchase this item. Need a web account? Register with us today!
17124221
View Documents
Cytiva
12085
100/Pk.
8500.00 €
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
Description
Description
  • High air flow rate venting applications
  • Offers broad chemical compatibility with PTFE membrane and polypropylene housing 100% integrity-tested to provide the high level of confidence you require
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
PTFE
Non-sterile
Stepped Hose Barb
0.2 μm
280 cm2
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.