missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 24mm PP Screw Cap, White, 24-400 Thread

Polypropylene caps are chemically inert and suitable for most chromatography applications.

Brand:  Fisherbrand™ 24 08 0592

12.36 EUR valid until 2025-12-31
Use promo code "24362" to get your promotional price.


 View more versions of this product

Product Code. 10759644

  • 17.40 EUR / Pack of 100
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Only null left
In Stock
Edge
Add to basket

Description

Description

  • 24mm PP Screw Cap
  • White
TRUSTED_SUSTAINABILITY
Specifications

Specifications

White
Polypropylene
Closed Top Screw Cap
24 mm
100
ND24
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.