Learn More
Buerkle™ White Disposable Forceps
BIOPHARMAGREENER_CHOICE
Disposable forceps are ideal for grasping, lifting, and moving small objects like fine tissue or solid samples.
Brand:  Buerkle™ 5378-1042
 View more versions of this product
Product Code. 16555210
  • 73.75 EUR / Pack of 100
Only null left
Estimated Shipment: 07-07-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • The fluted handles ensure optimum and secure handling as well as precise work
  • The light plastic forceps for single use allow for fatigue-free work
  • The clean room-manufactured lab assistants meet the highest standards and prevent cross-contamination or impurities in the sample being examined
  • The lab tweezers are suitable for chemical, pharmaceutical or cosmetic applications as well as the food industry and scientific uses
  • Sterile forceps are individually wrapped
  • For single use
  • Cleanroom manufactured (clean room class 7)
  • Complies with EU food requirements and FDA regulations
  • Pointed/angled tip: The disposable forceps have fine, smooth, curved gripping tips. Different materials can be held and processed reliably with the pointed ends
  • Pointed tip: The anatomical forceps have a straight tip, fluted on the inside. This makes it possible to grip sensitive materials reliably
  • Broad tip: The disposable forceps have ends with a broad tip. The broad tip is smooth and blunt. It prevents the material from being damaged when it is gripped
Specifications
Specifications
Broad
5.12 in.
130 mm
Sterile
100 (packaged individually)
Polystyrene
Forceps
Standard
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.