missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ CELLine™ Bioreactor Flask
Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric) Cell:
5 mL
15 mL
Capacity (Metric) Media:
350 mL
1000 mL
For Use With (Application):
Adherent Cultures
Suspension Cultures
No. per Case:
1/Cs.
3/Cs.
5/Cs.
Unit Size:
Each
Pack of 3
Pack of 5
Description
Includes
User manual with feeding and harvesting schedules
High-density cell culture flasks with dual membrane system allow for less culture maintenance and a high concentration harvest.
- Bulk media chamber allows for longer run times without media refreshing
- Cell compartment concentrates cells, antibodies and proteins for simplified purification
- For suspension cell culture and adherent cell culture: Hybridoma, CHO, NS0, SF, BHK, HEK, etc.
02-912-745, 02-912-742
- For adherent cell cultivation
- 1000 mL media compartment
- 15 mL cell compartment
- Available as package of one (02-912-745) or package three (02-912-742)
- For suspension cell cultivation
- 350 mL media compartment
- 5 mL cell compartment
- Available as package of one (02-912-746) or package five (02-912-746)
- For suspension cell cultivation
- 1000 mL media compartment
- 15 mL cell compartment
- Available as package of one (02-912-744) or package three (02-912-741)
Specifications
Specifications
| Capacity (English) Cell | 0.527 oz. |
| Capacity (English) Media | 35.2 oz. |
| Capacity (Metric) Cell | 15 mL |
| Capacity (Metric) Media | 1000 mL |
| For Use With (Application) | Suspension Cultures |
| Includes | User manual with feeding and harvesting schedules |
| No. per Case | 3/Cs. |
| Sterility | Sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction