missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Water, UHPLC-MS

Product Code. 15339865 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 L
Packaging:
Clear Borosilicate Glass Bottle
Unit Size:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer. | CAS: 7732-18-5 | H2O | 18.015 g/mol

Water, H2O, CAS Number-7732-18-5, dihydrogen oxide, dihydrogen monoxide, 1L, <-0.01 AU at 230nm, <-0.01 AU at 210nm, <-0.01 AU at 220nm, <-0.01 AU at 240nm, <-0.005 AU at 280nm, <-0.005 AU at 260nm, <-0.005 AU at 254nm, 2ppb max., 100 deg.C, 5ppb max., CHEBI:15377, Colorless, 1.000g/cm3, UHPLC-MS

TRUSTED_SUSTAINABILITY

Specifications

Absorbance ≤0.005 AU at 254nm, ≤0.005 AU at 260nm, ≤0.005 AU at 280nm, ≤0.01 AU at 210nm, ≤0.01 AU at 220nm, ≤0.01 AU at 230nm, ≤0.01 AU at 240nm
Chemical Name or Material Water
pH 7
CAS 7732-18-5
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Molecular Formula H2O
Synonym dihydrogen oxide, dihydrogen monoxide
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
PubChem CID 962
ChEBI CHEBI:15377
Grade UHPLC-MS
Packaging Clear Borosilicate Glass Bottle
Residue after Evaporation ≤1ppm
Vapor Pressure 17.5mmHg at 20°C
Density 1.000 g/cm3
Quantity 1 L
Physical Form Liquid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.