missing translation for 'onlineSavingsMsg'
Learn More

Water, BAKER HPLC ANALYZED, J.T. Baker™

Product Code. 10546602 Shop All J.T.Baker Products
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
Unit Size:
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

HPLC gradient grade water for use in Liquid Chromatography (HPLC & UHPLC) and Spectrophotometry, packaged in 2.5L glass bottles.

BAKER ANALYZED™ HPLC grade reagents are a group of high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations. They are optimized for Liquid Chromatography applications, including high performance liquid chromatography, gel permeation chromatography and UV-spectrophotometric analysis.

  • Manufactured and tested to assure interference-free separations
  • Highly characterized reagents that have unmatched lot-to-lot consistency
  • Minimizes recalibration or instrument adjustments
  • Solvents are controlled for high assay and low UV absorption, fluorescence, residue and water
  • An Actual Lot Analysis is provided on the label

Specifications

Absorbance Max. 0.005 au at 220nm, Max. 0.001 au at 254nm
pH 7
CAS 7732-18-5
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
Grade BAKER ANALYZED™ HPLC
Odor Odorless
Packaging Glass Bottle
Residue after Evaporation Max. 2ppm
Vapor Pressure 0.05 kPa
Density 1.00g/mL
Quantity 2.5 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.