missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Water with 0.05% Formic Acid, LC-MS Ultra, Honeywell™ Riedel-de-Haën™ CHROMASOLV™
Click to view available options
Quantity:
2 L
Packaging:
Glass Bottle
Unit Size:
2 L
Description
- For the most demanding LC-MS and UHPLC applications
- Tested for UHPLC-MS suitability
- Highest solvent purity for low detection limits
- Extremely low MS baseline noise
- Exemplary lot-to-lot reproducibility
- Low levels of trace metals
- Packaged in clear borosilicate glass bottles
Specifications
Specifications
| CAS | 7732-18-5, 64-18-6 |
| Quantity | 2 L |
| Packaging | Glass Bottle |
| Molecular Formula | H2O |
| UN Number | NONH for all modes of transport |
| Formula Weight | 18.02g/mol |
| Grade | LC-MS Ultra CHROMASOLV™ |
| Chemical Name or Material | Water With 0.05% Formic Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction