Learn More
IKA™ Vortex 2 Shaker
GREENER_CHOICE
Vortex shaker suitable for touch operation or continuous operation.
Brand:  IKA™ 0025000258
240.93 EUR valid until 2025-12-20
Use promo code "24121" to get your promotional price.
Product Code. 15523940
  • ONLINE EXCLUSIVE PRICE
    € 340.08
    € 302.00 / Each
    Save € 38.08
    11% Off
View Available Promotions
Only null left
In Stock
Edge
Add to basket
Description
Description
  • Touch function
  • Suitable for continuous operation (low heat due to ventilation of motor)
  • Stable operation at high speeds due to silicon base feet
  • Infinitely adjustable speed range
  • Various applications possible because of 3 interchangeable attachments and 7 inserts (e.g. Eppendorf tubes, microtiter plates, Erlenmeyer flasks 250 mL etc.), these need to be ordered separately
  • Attachments securely click onto appliance in any position
  • Special strap (VG 3.36) ensures easy handling of round/Erlenmeyer flasks
  • Sturdy metal die cast housing
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4 mm
220/240 V
EU Plug
138 x 120 x 140 mm
500 to 2500 rpm
60 W
50/60 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.