missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Vinyl Mini Pictograms Hazardous Labels

Product Code. 11313472
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Displayed Text:
CMR Mutagenic - Respiratory Risk
Combustible Material
Corrosive Material
DANGER for Aquatic Environment
DANGER of Cutaneous Sensitivity - inhalation
Explosive Material
Gases Under Pressure
Inflammable Solid Material
Toxic Material
missing translation for 'unitSize'
Pack of 96
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Displayed Text unitSize
11313472 CMR Mutagenic - Respiratory Risk Pack of 96
11353472 Combustible Material Pack of 96
11333472 Corrosive Material Pack of 96
12034128 DANGER for Aquatic Environment Pack of 96
11303472 DANGER of Cutaneous Sensitivity - inhalation Pack of 96
12024128 Gases Under Pressure Pack of 96
11333442 Explosive Material Pack of 96
11343442 Inflammable Solid Material Pack of 96
11363472 Toxic Material Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 missing translation for 'options'
Product Code. 11313472 missing translation for 'mfr' Brady™ missing translation for 'supplierNo' 814020

Please to purchase this item. Need a web account? Register with us today!

Edge

Specifications

Certifications/Compliance ISO 7010, RoHS compliant using EU Directive 2002/95/EC
Alternate Part Number MINI PIC1807-10X10-GHS08/96
Material Vinyl
Quantity 96/Pk.
Adhesive Type Permanent Acrylic
For Use With (Application) Pipe markers and safety signs
Height (Metric) 10 mm
Width (Metric) 200 mm
Temperature Range (Metric) -18°C to 60°C
Shape Diamond
Displayed Text CMR Mutagenic - Respiratory Risk
Product Type First Aid Sign
Color Black on White
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.