Learn More
3M™ Versaflo™ Intrinsically Safe Powered Air Turbo Starter Kit, TR-819E
Product Code. 16358981
Change view
Click to view available options
:
Product Type: PAPR System Kit, Color: Light Gray, Model: TR-819E
Product Code.
16358981 Product Type: PAPR System Kit, Color: Light Gray, Model: TR-819E
1 options
This item is not returnable. View return policy
Product Code. 16358981 Supplier 3M™ Supplier No. 7100169180
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
3M™ Versaflo™ Intrinsically Safe Powered Air Turbo Starter Kit TR-819E is a handy kit approved to Zone 0 requirements and may be suitable for areas where potentially explosive atmospheres are expected and permanent within normal operations. Ready-to-use and has everything you need to get started. Approved for use with a wide range of Versaflo™ Soft Headtops S Series and Rigid Headtops M Series. ATEX certified and CE approved.
  • Convenient ready-to-use kit
  • Approved for use with a wide range of Versaflo™ Soft Headtops S Series and Rigid Headtops M Series
  • Includes a rechargeable standard capacity lithium Ion battery
  • Headtop not included
  • CE approved
TRUSTED_SUSTAINABILITY
Specifications
Battery Type Lithium-ion
Certifications/Compliance CE Approved
Product Type PAPR System for Heavy Industry
Quantity 1/Cs.