missing translation for 'onlineSavingsMsg'
Learn More

Vancomycin Hydrochloride MP Biomedicals

Brand:  MP Biomedicals 0219554001


Quantity 1 g

Product Code. 11475492

  • 2435.00 DKK / 1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

123409-00-7
C66H257Cl2N9O24
C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.C.CC(C1=CC(=C(C=C1)OC2CC(C(C(O2)CO)O)O)OC3=CC=CC=C3)NC(=O)C.C(=O)O.N.N.N.N.N.N.N.N.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.Cl.Cl
1632.721
1 g
ARNOCQCQWLXLRG-UHFFFAOYSA-N
azane;N-[1-[4-[4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-3-phenoxyphenyl]ethyl]acetamide;formic acid;methane;pentadecahydrate;dihydrochloride
44134997
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Vancomycin Hydrochloride

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.