Learn More
Welch Ilmvac™ Vacuum Filtration And Desiccation Connection Kit
Facilitates proper connection to vacuum filtration or dissicator. Welch Ilmvac™ Vacuum Filtration And Desiccation Connection Kit includes a filter and suitable tubing.
Brand:  Welch Ilmvac™ 404008
Product Code. 13504920
  • ONLINE EXCLUSIVE PRICE
    176.00 €
    82.40 € / Each
    Save 93.60 €
    53% Off
Only null left
In Stock
Edge
Add to basket
Description
Description
  • Quick connection to vacuum filtration funnel / manifold or dessicator
  • Kit includes in-line hydrophobic filter, 1.5m silicone vacuum hose (6mm I.D.) and 1.5m silicone vacuum hose (8mm I.D.)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
In-Line Hydrophobic Filter, 1.5m Silicone Vacuum Hose (6mm ID) And 1.5m Silicone Vacuum Hose (8mm ID)
1 Kit
6 mm ID, 8 mm ID
Vacuum Filtration And Desiccation Connection Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.