Learn More
uvex i-3 Safety Goggles
Product Code. 15366062 Shop All Uvex Products
Change view
Click to view available options
Frame Color:
Anthracite Blue
Black, Light Gray
Lens Tint:
Clear
Gray
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Frame Color Lens Tint unitSize
15366062 Black, Light Gray Clear Each
12395793 Anthracite Blue Clear Each
15376062 Black, Light Gray Gray Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15366062 Supplier uvex Supplier No. 9190280
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy
Innovative metal-free protective spectacles, designed to perfectly fit the face
  • A new dimension in protective eyewear - these innovative metal-free protective spectacles are designed to perfectly fit the face
  • Ergonomically designed Softflex bridge piece, soft and variable nose loops do not slip and sit comfortably on the nose, Softflex ear pieces, variable side-arm
  • Angle, adjustable side-arm flex zone
  • Coating uvex supravision excellence
  • TRUSTED_SUSTAINABILITY
    Specifications
    Certifications/Compliance EN 170
    Lens Type Uvex™ supravision excellence
    Lens Coating Anti-fog
    Description Shield-lens spectacle
    Lens Tint Clear
    Product Type Ergonomic Safety Glasses
    Frame Color Black, Light Gray
    Material Polycarbonate
    Disposable Reusable
    Lens Material Polycarbonate
    Frame Material Plastic
    UV Protection UV400
    Frame Type Standard