missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uridine, ∽99%, MP Biomedicals™

39.25 EUR - 471.00 EUR
Chemical Identifiers
CAS | 58-96-8 |
---|---|
Molecular Formula | C9H12N2O6 |
Molecular Weight (g/mol) | 244.203 |
InChI Key | DRTQHJPVMGBUCF-DHRBYNEYSA-N |
Synonym | uridine, uridin, uracil riboside, 1-beta-d-ribofuranosyluracil, beta-uridine, d-uridine, b-uridine, uracil, 1-beta-d-ribofuranosyl, unii-whi7hq7h85, urd |
PubChem CID | 89495556 |
IUPAC Name | 1-[(3R,4S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
SMILES | C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11497431
![]() |
MP Biomedicals
0210321605 |
5 g |
39.25 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11407441
![]() |
MP Biomedicals
0210321610 |
10 g |
66.15 EUR
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11417441
![]() |
MP Biomedicals
0210321625 |
25 g |
162.00 EUR
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11427441
![]() |
MP Biomedicals
0210321680 |
100 g |
471.00 EUR
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
58-96-8 | |
244.203 | |
uridine, uridin, uracil riboside, 1-beta-d-ribofuranosyluracil, beta-uridine, d-uridine, b-uridine, uracil, 1-beta-d-ribofuranosyl, unii-whi7hq7h85, urd | |
1-[(3R,4S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione |
C9H12N2O6 | |
DRTQHJPVMGBUCF-DHRBYNEYSA-N | |
89495556 | |
C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O |
Specifications
58-96-8 | |
∽99% | |
uridine, uridin, uracil riboside, 1-beta-d-ribofuranosyluracil, beta-uridine, d-uridine, b-uridine, uracil, 1-beta-d-ribofuranosyl, unii-whi7hq7h85, urd | |
C1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O | |
244.203 | |
244.2 | |
Uridine |
White | |
C9H12N2O6 | |
DRTQHJPVMGBUCF-DHRBYNEYSA-N | |
1-[(3R,4S)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione | |
89495556 | |
Powder |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title