missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heidolph™ Unimax 1010
The Unimax 1010 is a medium sized lab shaker and accepts load capacities of 5 kg that is ideal in science, research, and other laboratory environments.
Brand: Heidolph™ 543-12310-00
1647.44 EUR valid until 2025-12-20
Use promo code "24121" to get your promotional price.
This item is not returnable.
View return policy
Description
- Shaker Heidolph Unimax 1010 platform size 290w x 258d mm, max speed 500rpm, max load 5kg, orbit 10mm, orbital shaking, includes timer and platform
- The Unimax 1010 is a medium sized lab shaker and accepts load capacities of 5 kg
- The shaking orbit of 10 mm performs a gentle motion for sensitive samples and supports culture plates and Erlenmeyer flasks
- Set and continuously adjust the variable speed on the digital display from 30 to 500 rpm
- This model is suitable for the modular incubator system and is recommended for applications which require variable temperature control up to 65° C
- An digital process timer allows for unattended operation and can be set from 1 to 999 minutes

Specifications
10 mm | |
30 to 500 rpm | |
Unimax 1010 |
5 kg | |
Shaker |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction