missing translation for 'onlineSavingsMsg'
Learn More

UBC13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1081.84 DKK - 3058.99 DKK

Specifications

Antigen UBC13
Concentration 0.22 mg/mL
Applications Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16836503
View Documents
Proteintech
10243-1-AP-20UL
20 μL
1081.84 DKK
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16826503
View Documents
Proteintech
10243-1-AP-150UL
150 μL
3058.99 DKK
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Anti-Ubc13 is a monoclonal antibody specific to the approximately19 kDa Ubc13 protein. Ubc13 is a ubiquitin conjugating enzyme that is complexed with a ubiquitin activating enzyme (E1), Mms2. The association of Mms2 to Ubc13 forms an active heterodimer that is involved in error-free post replicative DNA repair in yeast. Upon complexing with another conjugating enzyme variant, Uev1a, Ubc13 participates in the regulation of NF-kB signal transduction pathway in humans.
Specifications

Specifications

UBC13
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Mouse, Human, Rat
P61088, P61089, Q9EQX9
116725, 7334, 93765
UBC13 Fusion Protein Ag0292
Primary
-20°C
UBE2N
0.22 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
BLU, UBC13, UbcH ben, UBE2N, Ubiquitin carrier protein N, Ubiquitin protein ligase N
UBE2N
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.