Learn More
DuPont™ Tyvek 500 Laboratory Coat no pockets
Offers enhanced protection for body parts that are more exposed to hazardous substances. DuPont™ Tyvek™ 500 Lab Coat with Press Studs are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.
Merk: DuPont™ D13398884
Beschrijving
- Silicon non-added
- With 5 press stud closures
- Elasticated cuffs (not tunnelled)
- Stitched internal seams
- Permeable to both air and water vapour, yet repels water-based liquids and aerosols
- Excellent barrier against fine particles (down to 1 micron in size), is ultra-low-linting and antistatically treated
- Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
Specificatie
Large | |
White | |
Elastic | |
EN 1149-5 |
HDPE | |
Press Studs | |
Stitched (internal) |