Learn More
DuPont™ Tyvek™ IsoClean™ Cleanroom Sleeve
Durable and comfortable sleeve offering protection against dry particles and microorganisms.
Merk: DuPont™ D15531634
Beschrijving
- Tunnelled elastication at wrist and bicep
- Provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash
- Breathable and exceptionally low linting
- Accessory
Specificatie
Controlled Environment Sleeve | |
Non-sterile | |
IC501B00 | |
(EU) 2016/425, EN 14126 | |
Class 1000-100,000 (ISO 6-8) |
Polyethylene | |
Bound | |
Biotech, Pharmaceutical, Medical Device Manufacturing, Food Processing, Other Controlled Environments | |
45 cm |